Copper(II) sulphate pentahydrate USP

Supplier: AVANTOR PERFORMANCE MATERIALS US
Warning

Synonyms: Cupric sulphate pentahydrate, Copper sulphate pentahydrate

Management of Change (MOC) category = RL

1844-01 1844-05
JT1844-1CS 1853.28 USD
JT1844-1 JT1844-5
Copper(II) sulphate pentahydrate USP
Copper(II) sulphate pentahydrate
Formula: CuSO₄·5H₂O
MW: 249.69 g/mol
Density: 2.284 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149681
CAS Number: 7758-99-8
EINECS: 231-847-6
UN: 3077
ADR: 9 III
REACH: 01-2119520566-40
Merck Index: 13,02682

Order Now

Specification Test Results

Meets U.S.P Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Calcium (Ca) ≤0.005 %
USP - Identification Passes Test
USP - Iron (Fe) ≤0.003 %
USP - Loss on Drying 33.0 - 36.5 %
USP - Nickel (Ni) ≤0.005 %
USP - Potassium (K) ≤0.01 %
USP - Sodium (Na) ≤0.02 %
USP - Assay (CuSO₄) (dried basis) 98.5 - 100.5 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR