I-CHEM™ Vials with Screw Cap, Borosilicate Glass, Thermo Scientific
Supplier: Thermo Scientific
SB46-0040 C126-0020 C136-0040 C146-0040 C226-0020 C246-0040 C326-0020 C336-0040 C346-0040 S126-0020 S136-0040 S136-0060 S146-0040 S226-0020 S236-0040 S236-0060 S246-0040 S246-0060 S326-0020 S336-0040 S340-0120 S346-0040 S346-0060 SB36-0040 T136-0040 T236-0040 T336-0040 TB36-0040 C236-0040 S336-0060 T246-0040 T346-0040 C346-0020 S349-1000
46600-678CS 490.94 USD
46600-678 IRC126-0020 IRC136-0040 IRC146-0040 IRC226-0020 IRC246-0040 IRC326-0020 IRC336-0040 IRC346-0040 IRS126-0020 IRS136-0040 IRS136-0060 IRS146-0040 IRS146-0060 IRS226-0020 IRS236-0040 IRS236-0060 IRS246-0040 IRS246-0060 IRS326-0020 IRS336-0040 IRS340-0120 IRS346-0040 IRS346-0060 IRSB36-0040 IRT136-0040 IRT236-0040 IRT336-0040 IRTB36-0040 IRC236-0040 IRS336-0060 IRT246-0040 IRT346-0040 IRC346-0020 IRS349-1000
I-CHEM™ Vials with Screw Cap, Borosilicate Glass, Thermo Scientific
Vials
Thermo Scientific I-Chem Certified™ 200 and 300 series vials meet or exceed U.S
Thermo Scientific I-Chem 100 series vials are made from the same high-quality materials as Thermo Scientific IChem Certified 200 and 300 series, but are not processed and do not include a certificate or container labels. Certificates of Analysis are available by request for 200 series vials and economy packs, but unavailable for the 100 series vials.
EPA 'Specifications and Guidance for Contaminant-Free Sample Containers', and are provided with sample-analysis labels, unattached. Thermo Scientific I-ChemMore Product Information