I-CHEM™ Wide Mouth Jars, Amber Glass, with Caps, Thermo Scientific
Supplier: Thermo Scientific
341-1250 141-0060 141-0120 141-0250 141-0500 141-0950 241-0060 241-0120 241-0250 241-0500 241-0950 341-0060 341-0120 341-0500 341-0950 341-0250 141-2500 341-2500
13500-376CS 208.76 USD
13500-376 IR141-0060 IR141-0120 IR141-0250 IR141-0500 IR141-0950 IR241-0060 IR241-0120 IR241-0250 IR241-0500 IR241-0950 IR341-0060 IR341-0120 IR341-0500 IR341-0950 IRV341-0950 IR341-0250 IRV241-0950 IR141-2500 IR341-2500
I-CHEM™ Wide Mouth Jars, Amber Glass, with Caps, Thermo Scientific
Bottles Water Sampling Bottles
For use with UV-light sensitive samples.
  • With PTFE-lined white PP closed top cap (assembled on bottles)
  • Certified to meet or exceed EPA Performance Based Specifications for semi-volatile organics, pesticides, PCBs and metals analyses
  • Each jar individually barcoded with lot number and unique container number
I-CHEM Certified™ 200 and 300 series bottles are processed to meet or exceed US EPA Specifications and Guidance for Contaminent-Free Sample Containers. The 200 and 300 series jars include sample-analysis labels, unattached.
I-CHEM Certified™ 300More Product Information