1,1-Dimethylbiguanide hydrochloride 98%, white crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: AMPK Signaling Activator VII, Metformin, Metformin hydrochloride, 1,1-Dimethylbiguanide Hydrochloride

0215169101 0215169105 0215169110
IC15169101EA 55.12 USD
IC15169101 IC15169105 IC15169110
1,1-Dimethylbiguanide hydrochloride 98%, white crystalline powder
1,1-Dimethylbiguanide hydrochloride

1,1-Dimethylbiguanide hydrochloride is suitable biguanide agent used in a study to investigate its role in enhancing the secretions of plasma active glucagon-like peptide-1 (GLP-1) levels.


It may be used in the synthesis of the following:• bis(1,1-dimethylbiguanido)copper(II) octahydrate• bis(1,1-dimethylbiguanido)nickel(II)• 1,1-dimethylbiguanidium tetrabromothallate(III)


Metformin is an antidiabetic agent that reduces blood glucose levels and improves insulin sensitivity. Its metabolic effects, including the inhibition of hepatic gluconeogenesis, are mediated at least in part by activation of the LKB1-AMPK (AMP-activated protein kinase) pathway. Activation of this pathway also appears to be involved in the antiproliferative and proapoptotic actions of metformin in cancer cell lines.


Soluble in water (100 mg/ml - clear, colorless solution), 95% ethanol; insoluble in ether, acetone or chloroform.


Precaución: Research Use Only (RUO).

Formula: C₄H₁₂ClN₅
MW: 165.625 g/mol
MDL Number: MFCD00012582
CAS Number: 1115-70-4

Order Now

Specification Test Results

Identity Test Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR