Ammonium sulphate ≥99%, white crystalline powder, Enzyme Grade

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

0215037380 0215037390 0215037391
IC15037380EA 56.76 USD
IC15037380 IC15037390 IC15037391
Ammonium sulphate ≥99%, white crystalline powder, Enzyme Grade
Ammonium sulphate

Ammonium Sulfate may be used for the precipitation or fractionation of proteins or for purification of antibodies.


Useful for crystallographic analysis of nucleic acids and proteins. The use of ammonium sulfate in long PCR buffer, in PCR lysis solution, and in second strand cDNA synthesis has been reported.


Soluble freely in water; insoluble in alcohol, acetone.

Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
Merck Index: 13,00559

Order Now

Specification Test Results

Purity ≥99%
Heavy Metals ≤0,5 ppm (as Pb)
pH 5 - 6 (5% aq soln)
Protease, DNase, RNase Not detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR