Ethidium bromide, solution_dark red Molecular_Biology

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide

11ETBC1001
IC11ETBC1001EA 85.22 USD
IC11ETBC1001
Ethidium bromide, solution_dark red Molecular_Biology
Ethidium bromide

A 10 mg/mL Ready-To-Use solution of EtBr in specially filtered, deionized water.


  • Grade : Molecular Biology
  • Concentration : (10 mg/ml)
  • Suitability : Suitable for gel electrophoresis
  • Key Applications : Nucleic acid stain for PAGE or agarose gel electrophoresis; Used in fluorimetric assays for nucleic acids
  • Application Areas : Nucleic Acid Electrophoresis; Blotting & Detection; Nucleic Acid Gel Electrophoresis & Blotting
  • Product Type : Biochemicals
  • Analyte(s) Detected : dsDNA; dsRNA
  • Biochemical Category : Labels, Dyes & Stains
  • Chemical Class : Labels, Dyes & Stains
  • Vapor Pressure : 1.2 X 10-12 mm Hg at 25 °C
  • UV/Visible Absorbance : 254 nm
  • Presentation : Dark Red Solution
  • Detection Method : UV gel box (254 nm)
  • Solubility : Soluble in water.
  • Recommended Replacement For : Ethidium Bromide Powder
  • Certifications : DNase, RNase Protease Free
  • Detection/Incubation Time : 15-30 min with 0.5 mg/mL solution


Storage: protect from light.

Formula: C₂₁H₂₀BrN₃
MW: 394.31 g/mol
Boiling Pt: 72
Storage Temperature: Refrigerator
MDL Number: MFCD00011724
CAS Number: 1239-45-8
UN: 2810
Merck Index: 13,04751
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46d32fe41a7843db3a47bb81e3a7260969853086&vid=622319362092b4cd77673d1fd3bb941d1073357e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR