BioPulverizer™ System I, MP Biomedicals, LLC
Supplier: MP Biomedicals
116750200
IC116750200PK 146.12 USD
IC116750200
BioPulverizer™ System I, MP Biomedicals, LLC
Homogenizers Benchtop Homogenizers
BioPulverizer™ System I contains 10x2mL tubes of each of our most popular Lysing Matrix types (A, B, C, D and E).
  • Great starter pack for new FastPrep instrument owners
  • Optimize homogenization conditions
  • Contains 10 of each matrix: A, B, C, D and E
  • Impact-resistant 2 mL tubes
Matrix A, contains garnet matrix and one 1/4 inch ceramic sphere. Matrix B, contains 0.1 mm silica spheres. Matrix C contains 1 mm silica spheres. Matrix D, contains 1.4 mm ceramic spheres. Matrix E, Each impact-resistant 2 mL tube contains 1.4 ceramic spheres, 0.1 mm silicaMore Product Information