5-Fluoro-2'-deoxyuridine (floxuridine) ≥98%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: 5-Fluoro-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Fluoro-2'-deoxyuridine, Floxuridine

0210555110 0210555125 0210555183
IC10555110EA 57.92 USD
IC10555110 IC10555125 76102-654
5-Fluoro-2'-deoxyuridine (floxuridine) ≥98%, white powder
5-Fluoro-2'-deoxyuridine (floxuridine)

5-Fluorodeoxyuridine is an antineoplastic agent, which acts as an antimetabolite. When administrated by rapid injection it acts as flurouracil, but when infused slowly, usually intra-arterially, it is converted to active F-dUMP, which leads to enhanced inhibition of DNA synthesis. It inhibits DNA synthesis by blocking thymidylic acid synthetase.


  • Extinction Coefficient (E1%): EmM = 8.91 (0.1 M phosphate buffer, pH 6.0) (Lit.)
  • Optical Rotation: +37 ° (10 mg/mL in water at 25 °C) (Lit.)
  • pKa:7.6 (Lit.)


5-Fluorodeoxyuridine can be used to target thymidylate synthetase in cancer chemotherapy. Experimental anticancer agent shown to have activity against a variety of malignant neoplasms, including mouse mammary tumors and colorectal carcinomas.
Antineoplastic drug that acts as a potent inhibitor of thymidylate synthetase. Resistance to FUdR can develop in cancer cell cultures, among other means, by low-level Mycoplasma infection.


Soluble in water (clear, colourless solution); DMF.


Antineoplastic drug that acts as a potent inhibitor of thymidylate synthetase. Resistance to FUdR can develop in cancer cell cultures, among other means, by low-level Mycoplasma infection.


Precaución: Research Use Only (RUO).

Formula: C₉H₁₁FN₂O₅
MW: 246.19 g/mol
Melting Pt: 142…155 °C
Storage Temperature: Freezer
MDL Number: MFCD00006530
CAS Number: 50-91-9
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac12f5f394b06358f18bae318e5775a767221b68&vid=ac12f5f394b06358f18bae318e5775a767221b68&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard
Melting Point 142 - 155 °C
UV/VIS: l max (water) 266 - 270 nm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR