Sodium dodecyl sulphate (SDS) 99, powder_white

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

0210291880 0210291890 0210291891 0210291825
IC10291810EA 34.93 USD
IC10291810 IC10291880 IC10291890 IC10291891 76102-582
Sodium dodecyl sulphate (SDS) 99, powder_white
Sodium dodecyl sulphate (SDS)

SDS is an anionic detergent and wetting agent that is effective in both acid and alkaline solutions.


  • Auto Ignition: 248 °C (powder containing 90% SLS); 447 °C (solution containing 14.8%SLS) (Lit.)
  • Vapor Pressure: Practically zero (Lit.)
  • Aggregation Number: 62
  • Critical Micelle Concentration (CMC): 8.27mM
  • Soluble in water (200 mg/mL - clear, faint yellow solution), and ethanol (0.1 g/10 mL).


Most proteins bind SDS in a ratio of 1.4 grams SDS to 1 gram protein. The charges intrinsic to the protein become insignificant compared to the overall negative charge provided by the bound SDS. The charge to mass ratio is essentially the same for each protein and will migrate in the gel based only on protein size.
An anionic detergent typically used to solubilize and denature proteins for electrophoresis. SDS has also been used in large-scale phenol extraction of RNA to promote the dissociation of protein from nucleic acids when extracting from biological material.


SDS is an anionic detergent and wetting agent that is effective in both acid and alkaline solutions. Most proteins bind SDS in a ratio of 1.4 grams SDS to 1 gram protein. The charges intrinsic to the protein become insignificant compared to the overall negative charge provided by the bound SDS. The charge to mass ratio is essentially the same for each protein and will migrate in the gel based only on protein size. The critical micelle concentration (CMC) in pure water at 25 °C is 8.27 mM, and the aggregation number at this concentration is usually considered to be about 62. The micelle ionization fraction (α) is around 0.3 (or 30%).

Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Melting Pt: 180…210 °C
Density: 1.1 (20 °C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe7456afe8c507bd203020decd728221544bf8bc&vid=fe7456afe8c507bd203020decd728221544bf8bc&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
OD260 (1% aq soln) ≤0.1
OD280 (1% aq soln) ≤0.1
pH (1% aq soln) 6-7


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR