Butyric acid ≥99.5%, liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: Butanoic acid

0210116980
IC10116980EA 56.76 USD
IC10116980
Butyric acid ≥99.5%, liquid
Butyric acid

Butyric acid is used in the preparation of various butyrate esters. Low-molecular-weight esters of butyric acid, such as methyl butyrate, have mostly pleasant aromas or tastes. As a consequence, they find use as food and perfume additives. It is also used as an animal feed supplement, due to the ability to reduce pathogenic bacterial colonization.

Formula: C₄H₈O₂
MW: 88.11 g/mol
MDL Number: MFCD00002814
CAS Number: 107-92-6
UN: 2820
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR