Ham's F-10, MP Biomedical, LLC.
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
091040122
IC091040122EA 87.08 USD
IC091040122
Ham's F-10, MP Biomedical, LLC.
Cell Culture Media
Ham's F-10 Culture Media with L-glutamine and without sodium bicarbonate.
Originally, Ham's Nutrient Mixtures were developed for the clonal growth of several Chinese hamster ovary (CHO) clone cells, Hela cells and mouse L-cells. Both F-10 and F-12 are formulated for use with or without serum, depending on the type of cells being cultured. F-10 has demonstrated satisfactory growth of human diploid cells, white blood cells for chromosomal analysis and primary rat, rabbit and chicken tissue explants. Ham's F-12 is well suited for the growth of primary rat hepatocytes and prostate epithelial cells.
It is also the medium of choice for toxicity assays using CHO cells. MP also offers Ham's F-12 supplemented with 25 mM HEPES for improved buffering in the optimum pH range of 7.2 to 7.4. F12K is a modification of Ham's F-12 and Coon's F-12 with increased amino acid and pyruvate concentrations. Additionally, the salts have been modifed (Konigsbergs). F12K has been developed for culturing differentiated rat and chicken cells, as well as, primary human liver cells.