Uridine ≥99% cell culture reagent

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside

0219476301 0219476350
IC0219476301EA 71.6 USD
IC0219476301 IC0219476350
Uridine ≥99% cell culture reagent
Uridine

Uridine is a nucleoside, contains an uracil attached to a ribose ring (known as a ribofuranose) via a β-N1-glycosidic bond. It is an organic compound of the pyrimidine family, that occurs as one of the four bases coding genetic information in the polynucleotide chain of RNA.
Uridine can increase the rate-limiting step of Phosphatidylcholine synthesis in the brain, Phosphatidylcholine is the major structural and functional component of brain cell membranes. Without phosphatidylcholine, brain cells lose their youthful function and suffer the degenerative structural changes associated with aging. Uridine is thought to aid Alzheimer's Disease due to helping with synaptic connections, which appear to be reduced in the state of Alzheimer's.
Physical Appearance: White Powder
UV/Visible Absorbance: λ max (water)= 260 - 263 nm
Solubility: Soluble in 0.1 N HCl (50 mg/mL-clear, colorless solution) and water (50 mg/mL)

Formula: C₉H₁₂N₂O₆
MW: 244.20 g/mol
Melting Pt: 160…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00006526
CAS Number: 58-96-8
EINECS: 200-407-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c76c6cd4fec90024f42dbcfc139397f79025f107&vid=28610300dc95325ef2c7b6b5184595fc110998b8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR