Ac-Tyr-Val-Ala-Asp-AFC

Supplier: BACHEM AMERICAS INC

Synonyms: Ac-Tyr-Val-Ala-Asp-AFC

4027522.0005
I-1705.0005BAEA 392.15 USD
I-1705.0005BA
Ac-Tyr-Val-Ala-Asp-AFC
Ac-Tyr-Val-Ala-Asp-AFC

Sequence: Ac-Tyr-Val-Ala-Asp-AFC
Synonym(s): Ac-YVAD-AFC


The presence of halogen substituents at the fluorescent group improves membrane permeability of the YVAD-derived caspase-1 substrate.

Formula: C₂₂H₂₅F₂N₃O₇
MW: 481.453 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01318881
CAS Number: 219137-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR