Capping A THF/Pyridine/Acetic Anhydride (8/1/1), Novabiochem® for DNA/RNA synthesis
Supplier: MilliporeSigma
BI0222-0450 BI0222-4000
EM-BI0222-0450CS 956 USD
EM-BI0222-0450 EMBI0222-4000
Capping A THF/Pyridine/Acetic Anhydride (8/1/1), Novabiochem® for DNA/RNA synthesis
Capping Reagent
Capping Reagent A
  • Clear liquid form
  • Suitable for DNA, RNA and Oligonucleotide synthesis