4-(4-Nitrophenylazo)resorcinol for synthesis, Sigma-Aldrich®

Supplier: MERCK, S.A. DE C.V. SV

Synonyms: 4-((4-Nitrophenyl)diazenyl)benzene-1,3-diol, Azoviolet, Azo Violet, Magneson I

M-Clarity™ quality level = MQ200

8417130025
EM8.41713.0025CS 11012 USD
EM8.41713.0025
4-(4-Nitrophenylazo)resorcinol for synthesis, Sigma-Aldrich®
4-(4-Nitrophenylazo)resorcinol
Formula: C₁₂H₉N₃O₄
MW: 259.22 g/mol
MDL Number: MFCD00007310
CAS Number: 74-39-5
EINECS: 200-808-5

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 183 °C (decomposition)
Molar Mass 259.22 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR