Tween® 85 (Polysorbate) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Polyoxyethylenesorbitan Trioleate

M-Clarity™ quality level = MQ200

8.22188.0500
EM8.22188.0500CS 1308 USD
EM8.22188.0500
Tween® 85 (Polysorbate) for synthesis, Sigma-Aldrich®
Tween® 85 (Polysorbate)
Density: 1.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD01779641
CAS Number: 9005-70-3

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Vapor Pressure 1.4 hPa (20 °C)
Flash Point >149 °C DIN 51758
Density 1.03 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR