Tetraethylammonium tetrafluoroborate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Tetrylammonium tetrafluoroborate

M-Clarity™ quality level = MQ200

8.18246.0025 8.18246.0100
EM8.18246.0025CS 13203 USD
EM8.18246.0025 EM8.18246.0100
Tetraethylammonium tetrafluoroborate for synthesis, Sigma-Aldrich®
Tetraethylammonium tetrafluoroborate
Formula: C₈H₂₀BF₄N
MW: 217.06 g/mol
MDL Number: MFCD00011827
CAS Number: 429-06-1
UN: 2811

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 365 - 368 °C
Molar Mass 217.06 g/mol
Bulk Density 400 kg/m3
Hill Formula C8H20BF4N
Chemical Formula (C2H5)4NBF4


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR