(±)-1-Aminopropan-2-ol mixture of isomers ≥98% for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Aminopropanol, Hydroxypropylamine, 1-Aminopropan-2-ol, DL-1-Amino-2-propanol, (±)-Isopropanolamine, Aminoisopropyl alcohol, iso-propanol amine Amino-2-propanol DL-Isopropanolamine (±)-1-Amino-2-propanol (±)-Isopropanolamine

M-Clarity™ quality level = MQ200

8.07020.0100 8.07020.1000
EM8.07020.0100CS 757 USD
EM8.07020.0100 EM8.07020.1000
(±)-1-Aminopropan-2-ol mixture of isomers ≥98% for synthesis, Sigma-Aldrich®
(±)-1-Aminopropan-2-ol

Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Formula: C₃H₉NO
MW: 75.11 g/mol
Boiling Pt: 158…160 °C (1013 hPa)
Melting Pt: 1 °C
Density: 0.962 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008139
CAS Number: 78-96-6
EINECS: 201-162-7
UN: 2735
ADR: 8 II

Order Now

Specification Test Results

Assay isomers (GC, area%) ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C) 0.959 - 0.961
1-amino-2-propanol (GC, area%) ≥ 90.0 % (a/a)
Identity (IR) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR