Isoniazid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 4-Picolinic acid hydrazide, 4-Pyridinecarboxylic acid hydrazide, Isonicotinic hydrazide, Pyridine-4-carboxylic acid hydrazide, Isonicotinic acid hydrazide

M-Clarity™ quality level = MQ200

8.06753.0100
EM8.06753.0100CS 444 USD
EM8.06753.0100
Isoniazid for synthesis, Sigma-Aldrich®
Isoniazid
Formula: C₆H₇N₃O
MW: 137.14 g/mol
MDL Number: MFCD00006426
CAS Number: 54-85-3
Merck Index: 13,05205

Order Now

Specification Test Results

Solubility 125 g/l (20 °C)
Melting Point 169 - 172 °C
Molar Mass 137.14 g/mol
Bulk Density 800 kg/m3
Flash Point >250 °C
pH 6 - 8 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR