Sulfanilamide, GR for analysis, Supelco®

Supplier: MilliporeSigma

Synonyms: 4-Aminobenzenesulphonamide

M-Clarity™ quality level = MQ100

1.11799.0100
EM1.11799.0100CS 20861 USD
EM1.11799.0100
Sulfanilamide, GR for analysis, Supelco®
Sulphanilamide

1.11799.0100 is offered as an alternative to SX1150-2. Different Pack Size, Same Specifications.

Formula: H₂NC₆H₄SO₂NH₂
MW: 172.21 g/mol
Boiling Pt: 401 °C (1013 hPa)
Melting Pt: 163…167 °C
Density: 1.46 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007939
CAS Number: 63-74-1
EINECS: 200-563-4
Merck Index: 13,09010
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a76bb9ab6bdf23e9d846b24d4f57047dea1d5071&vid=f2f1c5e47a127cd33d3b4a2a31b1057c375456eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 7.5 g/l (25 °C)
Melting Point 163 - 167 °C
Molar Mass 172.2 g/mol
Bulk Density 500 kg/m3
Density 1.50 g/cm3
pH 5.8 - 6.1 (5 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR