Sodium hydroxide 0.01 M aqueous solution_concentrated, Titrisol®, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Caustic soda

1.09961.0001
EM1.09961.0001CS 1499 USD
EM1.09961.0001
Sodium hydroxide 0.01 M aqueous solution_concentrated, Titrisol®, Supelco®
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Density: 1.327 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8
Merck Index: 14,08627

Order Now

Specification Test Results

Solubility (20 °C) soluble
Density 1.01 g/cm3 (20 °C)
pH 13 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR