Water HPLC, Supelco®

Supplier: MilliporeSigma

Synonyms: Hydrogen oxide

WX0008-1
EM-WX0008-1CS 649 USD
EM-WX0008-1
Water HPLC, Supelco®
Water

(Hydrogen oxide)

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
UN: 1396
Merck Index: 13,10098

Order Now

Specification Test Results

Molar Mass 18.02
Appearance Clear liquid, free from particulates
Color (APHA) 10 max
Residue after evaporation 2 ppm max
Expiration date 12 months from manufacture date
Filtered Through 0.2 µm Filter To pass test
Fluorescence (as Quinine Base) 150 ppt max
Gradient at 210 nm 0.002 AU max
Gradient at 254 nm 0.0005 AU max
Odor To pass test
UV Abs. at 190 nm 0.01 AU max
UV Abs. at 210 nm 0.01 AU max
UV Abs. at 250 nm 0.005 AU max
UV Abs. at 280 nm 0.005 AU max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR