N,N-Dimethylformamide 99,9, OmniSolv®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF

DX1726P-1 DX1726-6 DX1726-1
EM-DX1726P-1CS 2427 USD
EM-DX1726P-1 EM-DX1726-6 EM-DX1726-1
N,N-Dimethylformamide 99,9, OmniSolv®, Supelco®
N,N-Dimethylformamide

For spectrophotometry and chromatography. UV cutoff 268 nm. Filtered.

Formula: HCON(CH₃)₂
MW: 73.09 g/mol
Boiling Pt: 153
Melting Pt: -61
Density: 0.949 g/cm³ (20 °C)
Flash Pt: 57.5 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3 III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.9% min
Color (APHA) 10 max
Expiration date 24 months
Filtered through 0.2 µm filter To pass test
Form Clear liquid
Infrared spectrum Conforms to standard
Residue after evaporation 5ppm max
UV Abs. at 268 nm 1.00AU max
UV Abs. at 270 nm 0.75AU max
UV Abs. at 275 nm 0.30AU max
UV Abs. at 295 nm 0.07AU max
UV Abs. at 310 nm 0.03AU max
UV Abs. at 340 nm 0.01AU max
UV Cut-off 268nm max
Water (H2O) 0.05% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR