Citric acid, anhydrous 99,5-100,5 alkalimetry on anhydrous substance, GR ACS ACS, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid

CX1723-7 CX1723-20 CX1723-3 CX1723-1
EM-CX1723-7EA 1054 USD
EM-CX1723-7 EM-CX1723-20 EM-CX1723-3 EM-CX1723-1
Citric acid, anhydrous 99,5-100,5 alkalimetry on anhydrous substance, GR ACS ACS, Supelco®
Citric acid

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: HOC(COOH)(CH₂COOH)₂
MW: 192.13 g/mol
Boiling Pt: 310 (dec.)
Melting Pt: 153…159 °C (lit.)
Density: 1.67 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011669
CAS Number: 77-92-9
EINECS: 201-069-1
REACH: 01-2119457026-42-0000
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d60f5a9b8e6c6dfb7eda04e1a1d5d6d971f92444&vid=caf4f9effc7fd22b942f7f92be770b06e40637a4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (alkalimetric on dried substance) 99.5-100.5%
Chloride (Cl) 0.001% max
Residue after ignition 0.02% max
Insoluble matter 0.005% max
Iron (Fe) 0.0003% max
Lead (Pb) 0.0002% max
Oxalate (C2O4) To pass test (<0.05%)
Phosphate (PO4) 0.001% max
Substances carbonized by hot sulfuric acid To pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR