TBE Buffer, Concentrate, OmniPur®. 10X.
Supplier: MilliporeSigma
8820-4L
EM-8830EA 718.88 USD
EM-8830 EM-8820
TBE Buffer, Concentrate, OmniPur®. 10X.
Buffers Electrophoresis Buffers Running Buffers
Liquid
Each liter of concentrate makes 10L of 1X TBE (0.089M TRIS, 0.089M Borate, 0.002M EDTA). DNase-, RNase-, and protease-free.