VWR® Contour™ Flammable Liquid Fume Hood Cabinets
Supplier: VWR International
CFG-2685-A2 CFG-2686-A2 CFG-2688-A2 CFG-2684-V CFG-2684-VL CFG-2685-V CFG-2686-V CFG-2688-V CWG-2685-A2 CWG-2686-A2 CWG-2688-A2
CFG-2685-A2EA 4018.6 USD
CFG-2685-A2 CFG-2686-A2 CFG-2688-A2 CFG-2684-V CFG-2684-VL CFG-2685-V CFG-2686-V CFG-2688-V CWG-2685-A2 CWG-2686-A2 CWG-2688-A2
VWR® Contour™ Flammable Liquid Fume Hood Cabinets
Furniture Storage Cabinets
Flammable storage cabinets are specifically designed for the storage of flammable and combustible liquids.
  • Fume hood flammable liquid base unit
  • One or two doors
  • Solvent storage
Both steel and wood cabinets meet UFC, OSHA and NFPA No. 30-1993 construction standards and are UL listed. A 2" deep, steel, liquid-tight pan covers the entire bottom to contain liquid leaks and spills. A second pan is provided as a full-depth adjustable shelf. Two diametrically opposed vents with spark screens are provided in the back for cases when ventilation is required.More Product Information