L(+)-Histidine ≥98%

Supplier: BeanTown Chemical

Synonyms: L-3-Imidazol-4-ylalanine, (S)-(+)-Histidine, (S)-2-Amino-3-(4-imidazolyl)propionic acid, H-His-OH, NSC 137773, L-Histidine

222780-100G 222780-25G 222780-500G
BT222780-100GEA 88.09 USD
BT222780-100G BT222780-25G BT222780-500G
L(+)-Histidine ≥98%
L(+)-Histidine

CAS: 71-00-1; EC No: 200-745-3; MDL No: MFCD00064315; RTECS: MS3070000 Solid; Molecular Formula: C6H9N3O2; MW: 155.15 Melting Point: 282° (decomposes) Optical Rotation: [α]20/D -39±1°, c = 2% in water


  • RTECS: MS3070000
  • Optical rotation: [α]20/D -39±1°, c = 2% in water
  • Solid

Formula: C₆H₉N₃O₂
MW: 155.16 g/mol
Melting Pt: 282 °C (dec.)
MDL Number: MFCD00064315
CAS Number: 71-00-1
EINECS: 200-745-3
Merck Index: 13,04741
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=badfdb36aac0611e5823a7d05e6cba944f6650fa&vid=9e9f9d881a5b3314c7f342312e77e13493901200&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR