Vanadium(V) oxide ≥99.9%

Supplier: BeanTown Chemical

Synonyms: Vanadium pentoxide, di-Vanadium pentaoxide

216910-100G 216910-25G
BT216910-100GEA 1054 USD
BT216910-100G BT216910-25G
Vanadium(V) oxide ≥99.9%
Vanadium(V) oxide

CAS: 1314-62-1; EC No: 215-239-8; MDL No: MFCD00011457; RTECS: YW2125000 UN No: UN2862; Haz Class: 6.1; Packing Group: III 3-12mm Pieces; Linear Formula: V2O5; MW: 181.88 Melting Point: 690°; Boiling Point: 1750° (decomposes) Density (g/mL): 3.357


  • RTECS: YW2125000
  • 3-12mm Pieces

Formula: O₅V₂
MW: 181.88 g/mol
Boiling Pt: 1750 (dec.)
Melting Pt: 690
Density: 3.357 g/mL
MDL Number: MFCD00011457
CAS Number: 1314-62-1
EINECS: 215-239-8
UN: 2862
ADR: 6.1 III
Merck Index: 13,09987

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR