Sodium gluconate ≥97%

Supplier: BeanTown Chemical

Synonyms: D-Gluconic acid sodium salt, Sodium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate, Gluconic acid, sodium salt, Sodium D-gluconate

215720-10KG 215720-2.5KG
BT215720-10KGEA 314 USD
BT215720-10KG BT215720-2.5KG
Sodium gluconate ≥97%
Sodium gluconate

CAS: 527-07-1; EC No: 208-407-7; MDL No: MFCD00064210; RTECS: LZ5235000 Solid; Molecular Formula: C6H11NaO7; MW: 218.14 Melting Point: 170-175°


  • RTECS: LZ5235000
  • Solid

Formula: HOH₂C(CH(OH))₄COONa
MW: 218.14 g/mol
Melting Pt: 170…175 °C
MDL Number: MFCD00064210
CAS Number: 527-07-1
EINECS: 208-407-7
Merck Index: 12,08766
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eafd5b786cb23fb5c20dd64ad056ed2d80dc0186&vid=b1ffe48dafca4b8bd9b0d709ea3e986099051767&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR