(S)-(-)-α-Methylbenzylamine ≥99%, ee 99% ee

Supplier: BeanTown Chemical

Synonyms: (S)-(-)-1-Phenylethylamine, (S)-(-)-α-Phenylethylamine, L(-)-α-Methylbenzylamine

214065-100G 214065-25G 214065-5G
BT214065-100GEA 158.63 USD
BT214065-100G BT214065-25G BT214065-5G
(S)-(-)-α-Methylbenzylamine ≥99%, ee 99% ee
(S)-(-)-α-Methylbenzylamine

CAS: 2627-86-3; EC No: 220-098-0; MDL No: MFCD00064406 UN No: UN2735; Haz Class: 8; Packing Group: III Liquid; Molecular Formula: C8H11N; MW: 121.18 Boiling Point: 187°; Flash point: 70°C (158°F) Density (g/mL): 0.94; Refractive Index: 1.526; Optical Rotation: [α]20/D -39°, neat Air Sensitive


  • Optical rotation: [α]20/D -39°, neat
  • Refractive index: 1.526
  • Air Sensitive
  • Liquid

Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 187 °C
Melting Pt: -10
Density: 0.94 g/mL
Flash Pt: 70 °C (158 °F)
MDL Number: MFCD00064406
CAS Number: 2627-86-3
EINECS: 220-098-0
ADR: 8 III
Merck Index: 12,06106

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR