Tetraethylammonium fluoride hydrate ≥97%

Supplier: BeanTown Chemical

Synonyms: Tetrylammonium fluoride hydrate, Tetraethylammonium fluoride xhydrate

212705-100G 212705-25G 212705-5G
BT212705-100GEA 388.16 USD
BT212705-100G BT212705-25G BT212705-5G
Tetraethylammonium fluoride hydrate ≥97%
Tetraethylammonium fluoride hydrate

CAS: 98330-04-2; EC No: 211-559-7; MDL No: MFCD00149990 Crystalline; Linear Formula: (CH3CH2)4N(F)·xH2O; MW: 149.26 (anhydrous) Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: C₈H₂₀FN·xH₂O
MW: 149.25 g/mol (anhy)
MDL Number: MFCD00149990
CAS Number: 98330-04-2
EINECS: 211-559-7

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR