Iron(III) phosphate hydrate

Supplier: BeanTown Chemical

Synonyms: Iron phosphate hydrate, Iron (III) orthophosphate hydrate

140565-1KG 140565-250G 140565-5KG
BT140565-1KGEA 95 USD
BT140565-1KG BT140565-250G BT140565-5KG
Iron(III) phosphate hydrate
Iron(III) phosphate hydrate

CAS: 51833-68-2; EC No: 233-149-7; MDL No: MFCD00016096 Powder; Linear Formula: FePO4·xH2O; MW: 150.82 (anhydrous)


  • Powder

Formula: FeO₄P·xH₂O
MW: 168.83
MDL Number: MFCD00016096
CAS Number: 51833-68-2
EINECS: 233-149-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR