Ruthenium ≥99.9% (trace metals basis), cubes 6 mm square

Supplier: BeanTown Chemical

140355-1PC
BT140355-1PCEA 968 USD
BT140355-1PC
Ruthenium ≥99.9% (trace metals basis), cubes 6 mm square
Ruthenium

CAS: 7440-18-8; EC No: 231-127-1; MDL No: MFCD00011207 Solid; Molecular Formula: Ru; MW: 101.07 Melting Point: 2310°; Boiling Point: 3900° Density (g/mL): 12.45


  • Solid

Formula: Ru
MW: 101.07
Boiling Pt: 3900 °C
Melting Pt: 2310 °C
Density: 12.45 g/mL
MDL Number: MFCD00011207
CAS Number: 7440-18-8
EINECS: 231-127-1

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR