Gallium trinitrate hydrate ≥99.999% (trace metals basis)

Supplier: BeanTown Chemical

138930-100G 138930-25G 138930-5G
BT138930-100GEA 809 USD
BT138930-100G BT138930-25G BT138930-5G
Gallium trinitrate hydrate ≥99.999% (trace metals basis)
Gallium trinitrate hydrate

CAS: 69365-72-6; EC No: 236-815-5; MDL No: MFCD00149723 UN No: UN1477; Haz Class: 5.1; Packing Group: III Crystalline Aggregates; Linear Formula: Ga(NO3)3·xH2O; MW: 255.73 (anhydrous) Melting Point: 110° (decomposes) Hygroscopic


  • Hygroscopic
  • Crystalline Aggregates

Formula: GaN₃O₉·H₂O
MW: 273.75
Melting Pt: 110 °C (dec.)
MDL Number: MFCD00149723
CAS Number: 69365-72-6
EINECS: 236-815-5
UN: 1477
ADR: 5.1 III

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR