Lead(II) oxide 99,9 Trace metals basis

Supplier: BeanTown Chemical

Synonyms: Lead monoxide

130700-2KG 130700-500G
BT130700-2KGEA 76 USD
BT130700-2KG BT130700-500G
Lead(II) oxide 99,9 Trace metals basis
Lead(II) oxide

CAS: 1317-36-8; EC No: 215-267-0; MDL No: MFCD00011164; RTECS: OG1750000 UN No: UN3077; Haz Class: 9; Packing Group: III -325 Mesh Powder; Linear Formula: PbO; MW: 223.19 Melting Point: 886°


  • RTECS: OG1750000
  • -325 Mesh Powder

Formula: PbO
MW: 223.2 g/mol
Boiling Pt: 1470
Melting Pt: 888
MDL Number: MFCD00011164
CAS Number: 1317-36-8
EINECS: 215-267-0
UN: 3077
ADR: 6.1 III
Merck Index: 13,05431

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR