Ruthenium(III) chloride oxide

Supplier: BeanTown Chemical

Synonyms: Ruthenium oxychloride, Tetraamminechlorohydroxoruthenium(III) chloride, Ruthenium Red

128530-1G 128530-5G
BT128530-1GEA 95 USD
BT128530-1G BT128530-5G
Ruthenium(III) chloride oxide
Ruthenium(III) chloride oxide

CAS: 11103-72-3; MDL No: MFCD00011479; RTECS: VM2656500 Crystalline; Linear Formula: [(NH3)5RuORu(NH3)4ORu(NH3)5]Cl6; Molecular Formula: H42Cl6N14O2Ru3; MW: 786.35 Melting Point: 500° (decomposes) Density (g/mL): 3.11


  • RTECS: VM2656500
  • Crystalline

Formula: H₄₂Cl₆N₁₄O₂Ru₃
MW: 786.354 g/mol
Melting Pt: >500 °C (dec.)
Density: 3.11 g/mL
MDL Number: MFCD00011479
CAS Number: 11103-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR