2-Piperazinone ≥97%

Supplier: BeanTown Chemical

Synonyms: Piperazin-2-one

127280-1G 127280-5G
BT127280-1GEA 83 USD
BT127280-1G BT127280-5G
2-Piperazinone ≥97%
2-Piperazinone

CAS: 5625-67-2; MDL No: MFCD01318687 UN No: UN1759; Haz Class: 8; Packing Group: III Powder; Molecular Formula: C4H8N2O; MW: 100.12 Melting Point: 136-140° Hygroscopic


  • Hygroscopic
  • Powder

Formula: C₄H₈N₂O
MW: 100.12 g/mol
Melting Pt: 136…140 °C
MDL Number: MFCD01318687
CAS Number: 5625-67-2
ADR: 8 III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR