Graphite (natural) ≥99%, crystalline powder -300 mesh

Supplier: BeanTown Chemical

125640-250G
BT125640-1KGEA 75.97 USD
BT125640-1KG BT125640-250G
Graphite (natural) ≥99%, crystalline powder -300 mesh
Graphite

CAS: 7782-42-5; EC No: 231-955-3; MDL No: MFCD00144065 UN No: UN1325; Haz Class: 4.1; Packing Group: III Powder; Molecular Formula: C; MW: 12.011


  • Powder

Formula: C
MW: 12.01 g/mol
Melting Pt: 3827
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-955-3
ADR: 4.1 III
Merck Index: 12,04560

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR