Mercury(II) trifluoromethanesulfonate ≥98%

Supplier: BeanTown Chemical

Synonyms: Mercury(II) trifluoromethanesulphonate, Mercury triflate

124540-1G 124540-25G 124540-5G
BT124540-1GEA 44 USD
BT124540-1G BT124540-25G BT124540-5G
Mercury(II) trifluoromethanesulfonate ≥98%
Mercury(II) trifluoromethanesulphonate

CAS: 49540-00-3; MDL No: MFCD00144746 UN No: UN2025; Haz Class: 6.1; Packing Group: II Powder; Linear Formula: Hg(O3SCF3)2; Molecular Formula: C2F6HgO6S2 ; MW: 498.72 Melting Point: 350° Moisture Sensitive


  • Moisture Sensitive
  • Powder

Formula: C₂F₆HgO₆S₂
MW: 498.73
Melting Pt: >350 °C
MDL Number: MFCD00144746
CAS Number: 49540-00-3
UN: 2025
ADR: 6.1 II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR