J-Lite™ JLA-9.1000 Fixed-Angle Rotors, Beckman Coulter®
Supplier: Beckman Coulter
366754
BK366754EA 22629.49 USD
BK366754 BK969331 BK969330
J-Lite™ JLA-9.1000 Fixed-Angle Rotors, Beckman Coulter®
Centrifuges Rotors
Lightweight, high-performance, four-place, aluminum, and fixed-angle rotors are for use in Avanti™ J-26 XP, XPI, and J-E centrifuges.
  • Ideal for General-Purpose, Large-Volume Applications Such as Pelleting Of Bacteria, Subcellular Organelles, Viruses, and Precipitates
  • Space for Four 1000 mL Bottles
  • Lightweight
  • High-Performance
  • Material: Aluminum
They are ideal for general-purpose, large-volume applications such as pelleting of bacteria, subcellular organelles, viruses, and precipitates.
They accommodate four 1000 mL bottles.
Información paraMore Product Information