2,2,4-Trimethylpentane ≥99.75% PRF, B&J Brand™ ASTM D2699/D2700, Burdick & Jackson™

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

Synonyms: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane

RS362-19 RS362-204
BJRS362-19EA 2779 USD
BJRS362-19 BJRS362-204
2,2,4-Trimethylpentane ≥99.75% PRF, B&J Brand™ ASTM D2699/D2700, Burdick & Jackson™
2,2,4-Trimethylpentane

Primary Reference Fuels


  • Octane number, the principal performance characteristic of gasoline, is determined through use of certified PRF fuels and a Cooperative Fuel Research (CFR) engine under controlled test conditions
  • used in both research and analytical applications
  • thorough quality control testing to ensure each lot conforms to the specifications defined in ASTM D2699 for Motor Octane Number (MON) and ASTM 2700 for Research Octane Number (RON).


Our ISO 9001 and 14001 certified research and manufacturing facility located in Muskegon, Michigan benefits from the Honeywell Operating System, a comprehensive and integrated manufacturing approach based on Six Sigma and Lean Manufacturing methodologies, ensuring unparalleled batch-to-batch consistency.


Certificaciones: Honeywell certifies that this product meets the requirements as defined in ASTM D2699 and D2700.

Formula: (CH₃)₂CHCH₂C(CH₃)₃
MW: 114.23 g/mol
Boiling Pt: 99.2
Melting Pt: -107
Density: 0.69 g/cm³ (20 °C)
Flash Pt: –8 °C
Storage Temperature: Ambient
MDL Number: MFCD00008943
CAS Number: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3 II
Merck Index: 13,05212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=bb9d5eb96909b6bebc6a8709db78c4ea0ef52430&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless liquid
Color (APHA) < 10
Water (Karl Fischer) < 0.010%
Residue < 5 mg/L
GC Purity > 99.75%
n-Heptane < 0.1%
Lead Content < 0.5 mg/L
Sulfur Compounds (as S) < 0.005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR