N-Methyl-2-pyrrolidone (NMP) ≥99.5%, BioSyn® for biochemistry, for applications requiring low water content, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, 1-Methyl-2-pyrrolidone, NMP, Methyl Pyrrolidone

BC305-200
BJBB305-4CS 3117.51 USD
BJBB305-4 BJBC305-200
N-Methyl-2-pyrrolidone (NMP) ≥99.5%, BioSyn® for biochemistry, for applications requiring low water content, Burdick & Jackson™
N-Methyl-2-pyrrolidone (NMP)

BioSyn« products are ideal for DNA, RNA and oligonucleotide synthesis. All solvents and regents are filtered (0.2-micron) and tested for oxidation reage.


  • Ultra-low water specifications
  • Low amine specifications
  • Low particulate levels
  • High solvent purity

Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 202
Melting Pt: -24
Flash Pt: 91
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Amines (as dimethylamine) 3 ppm max.
Water by Karl Fischer Titration 200 ppm max.
GC Analysis 99.5 % min.
Refractive Index (20ºC) 1.4680 - 1.4720
Residue 5 mg/L max.
UV Absorbance @ 300 nm 0.5 AU max.
UV Absorbance @ 325 nm 0.1 AU max.
UV Absorbance @ 350 nm 0.03 AU max.
UV Absorbance @ 400 nm 0.01 AU max.
UV Cutoff 285 nm max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR