Squalane 99,0 GC_ASSAY_METHOD GC, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: 2,6,10,15,19,23-Hexamethyltetracosane

85629-50ML
BJ85629-50MLEA 142 USD
BJ85629-50ML
Squalane 99,0 GC_ASSAY_METHOD GC, Fluka™
Squalane
Formula: C₃₀H₆₂
MW: 422.82 g/mol
Boiling Pt: 470
Melting Pt: -35
Density: 0.82 g/cm³ (20 °C)
Flash Pt: 218 °C
Storage Temperature: Ambient
MDL Number: MFCD00008953
CAS Number: 111-01-3
Merck Index: 12,08923

Order Now

Specification Test Results

Identity (IR) complying
Identity (NMR) complying
Assay (GC) Min. 99.0 %
Refractive index (n 20/D) 1.451 - 1.453


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR