Disodium hydrogen phosphate, anhydrous 99,0 Titrimetric, Puriss. p.a. ACS, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: sec-Sodium phosphate, di-Sodium hydrogen orthophosphate, di-Sodium hydrogen phosphate anhydrous, Sodium hydrogen phosphate, Disodium phosphate, Sodium phosphate dibasic

71640-1KG 71640-250G 71640-5KG
BJ71640-1KGEA 147.04 USD
BJ71640-1KG BJ71640-250G BJ71640-5KG
Disodium hydrogen phosphate, anhydrous 99,0 Titrimetric, Puriss. p.a. ACS, Fluka™
Disodium hydrogen phosphate
Formula: Na₂HPO₄
MW: 141.96 g/mol
Melting Pt: 250
Storage Temperature: Ambient
MDL Number: MFCD00003496
CAS Number: 7558-79-4
EINECS: 231-448-7
Merck Index: 12,08805

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
Colour of the solution complying
Clearness of the solution complying
pH (5 %, 25 °C) 8.7 - 9.3
Loss on drying (105°C) Max. 0.2 %
Water insoluble matter Max. 0.01 %
Arsenic (As) Max. 1 ppm
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 10 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 2 ppm
Zinc (Zn) Max. 5 ppm
Heavy metals (as Pb) Max. 10 ppm
Total S (as SO₄) Max. 50 ppm
Chloride (Cl) Max. 10 ppm
Total N Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR