Chloroform, Hydranal™ solubilizer for Karl Fischer titration, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

37863-6X1L
BJ37863-1LCS 507 USD
BJ37863-1L
Chloroform, Hydranal™ solubilizer for Karl Fischer titration, Fluka™
Chloroform
Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 61.7
Melting Pt: -63
Density: 1.48 g/cm³ (20 °C)
Storage Temperature: Room Temperature
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 3 III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Suitability (for KF-titration) test passed
Assay of water Max. 0.01 %
Number Of Samples Min. 2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR