HYDRANAL® Karl Fischer Reagents, Honeywell Research Chemicals
Supplier: Honeywell Research Chemicals
34816-6X1L-US 34730-6X1L 34730-4X2.5L 34732-6X500ML 34734-6X1L 34734-4X2.5L 34697-6X1L 34697-4X2.5L 34738-500ML 34749-6X1L 37855-6X1L 37856-6X1L 34805-6X1L-US 34805-4X2.5L-US 34805-6X500ML-US 34806-6X1L-US 34806-4X2.5L-US 34806-6X500ML-US 34816-4X2.5L-US 34816-6X500ML-US 34827-6X1L-US 34827-6X500ML-US 34698-6X1L-US 34723-6X1L-US 34738-6X1L-US 34800-6X1L-US 34800-4X2.5L-US 34801-6X1L-US 34801-4X2.5L-US 34801-6X500ML-US 34811-6X1L-US 34811-6X500ML-US 34812-6X1L-US 34812-4X2.5L-US 34817-4X1L-US 37817-6X1L-US 37817-4X2.5L-US 34732-500ML 34730-2.5L 34730-6X500ML
BJ34698-1LCS 801.43 USD
BJ34698-1L BJ34800-1L BJ34801-1L BJ34806-1L BJ34811-1L BJ34816-1L BJ37817-1L BJ34817-1L BJ34723-1L BJ34730-1L BJ34730-2.5L BJ34730-500ML BJ34732-500ML BJ34734-1L BJ34734-2.5L BJ34697-1L BJ34697-2.5L BJ34738-1L BJ34738-500ML BJ34749-1L BJ37855-1L BJ37856-1L BJ34805-1L-US BJ34805-2.5LUS BJ34805500MLUS BJ34806-1L-US BJ34806-2.5LUS BJ34806500MLUS BJ34816-1L-US BJ34816-2.5LUS BJ34816500MLUS BJ34827-1L-US BJ34827500MLUS BJ34698-1L-US BJ34723-1L-US BJ34732100MLUS BJ34738-1L-US BJ34800-1L-US BJ34800-2.5LUS BJ34801-1L-US BJ34801-2.5LUS BJ34801500MLUS BJ34811-1L-US BJ34811500MLUS BJ34812-1L-US BJ34812-2.5LUS BJ34817-4X1LUS BJ37817-1L-US BJ37817-2.5LUS BJ-34732-500ML BJ-34730-2.5L BJ34730-6X500ML
HYDRANAL® Karl Fischer Reagents, Honeywell Research Chemicals
Karl Fischer Titrants and Reagents
Original and Innovative Karl Fischer Reagents to analyze or measure water content
Our premium Hydranal™ product range for pyridine-free and ethanol-based water determination by Karl Fischer titration (KF titration) is comprised of one- and two-component reagents for volumetric Karl Fischer titrations. Also available are water standards for titer standardization, pH buffering reagents, special reagents for aldehydes, ketones, and other solubilizing agents.
Order Now
SPECIFICATIONS
Environmentally Preferable