Potassium chromate ≥99.5%, Puriss. p.a. Reag. Ph. Eur., ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger

31250-250G 31250-1KG
BJ31250-250GEA 133.55 USD
BJ31250-250G BJ-31250-1KG
Potassium chromate ≥99.5%, Puriss. p.a. Reag. Ph. Eur., ACS, Fluka™
Potassium chromate
Formula: K₂CrO₄
MW: 194.19 g/mol
Boiling Pt: 1000
Melting Pt: 985
Density: 2.732 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011368
CAS Number: 7789-00-6
EINECS: 232-140-5
UN: 3288
ADR: 6.1 III
Merck Index: 12,07784

Order Now

Specification Test Results

Assay Min. 99.5 %
Water insoluble matter Max. 0.005 %
pH (5 %, 25 °C) 8.6 - 9.8
Calcium (Ca) Max. 0.005 %
Iron (Fe) Max. 0.002 %
Sodium (Na) Max. 0.02 %
Lead (Pb) Max. 0.005 %
Chloride (Cl) Max. 0.001 %
Sulfate (SO₄) Max. 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR