Zinc acetate dihydrate 99-102, Puriss., meetSpec_DAB,E650, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Acetic acid zinc salt dihydrate, Zinc (II) acetate dihydrate, Zinc diacetate dihydrate

25044-1KG
89428-898EA 493.95 USD
89428-898 BJ25044-1KG
Zinc acetate dihydrate 99-102, Puriss., meetSpec_DAB,E650, Fluka™
Zinc acetate dihydrate
Formula: (H₃CCOO)₂Zn·2H₂O
MW: 219.51 g/mol
Melting Pt: 237
Density: 1.750 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066961
CAS Number: 5970-45-6
EINECS: 209-170-2
Merck Index: 12,10256

Order Now

Specification Test Results

Assay 99.0 - 102.0 %
pH (5 %, 25 °C) 6.0 - 8.0
Water insoluble matter Max. 0.005 %
in CH₃COOH insoluble matters Max. 0.005 %
Alkalis/earth alkalis Max. 0.2 %
Arsenic (As) Max. 3 ppm
Calcium (Ca) Max. 0.005 %
Cadmium (Cd) Max. 5 ppm
Iron (Fe) Max. 50 ppm
Potassium (K) Max. 0.005 %
Magnesium (Mg) Max. 0.005 %
Sodium (Na) Max. 0.005 %
Lead (Pb) Max. 20 ppm
Chloride (Cl) Max. 50 ppm
Sulfate (SO₄) Max. 100 ppm
Organic volatile impurities (GC) complying
Residual solvents complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR