Iron(III) chloride hexahydrate 97% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Iron trichloride hexahydrate, Ferric chloride hexahydrate

236489-100G 236489-500G 236489-6X500G
BJ236489-100GEA 97 USD
BJ236489-100G BJ236489-500G BJ2364896X500G
Iron(III) chloride hexahydrate 97% ACS, Fluka™
Iron(III) chloride hexahydrate
Formula: FeCl₃·6H₂O
MW: 270.3 g/mol
Boiling Pt: 280…285 °C
Melting Pt: 37 °C
Density: 2.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149712
CAS Number: 10025-77-1
UN: 3260
ADR: 8 III
Merck Index: 13,04048
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=808c75376e55f29296d58eb1e5a1e184039ce733&vid=6d820873011bee3ff401441544f7f85599e40626&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 97.0 - 102.0 %
Insoluble in acid Max. 0.01 %
Calcium (Ca) Max. 0.01 %
Copper (Cu) Max. 0.003 %
Potassium (K) Max. 0.005 %
Magnesium (Mg) Max. 0.005 %
Sodium (Na) Max. 0.05 %
Zinc (Zn) Max. 0.003 %
Iron (Fe 2⁺) Max. 0.002 %
Nitrate (NO₃) Max. 0.01 %
Total phosphorus (as PO₄) Max. 0.01 %
Sulfate (SO₄) Max. 0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR