Magnesium perchlorate ≥85% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Anhydrone

222283-500G
BJ222283-100GEA 105.92 USD
BJ222283-100G BJ222283-500G BJ222283-5G
Magnesium perchlorate ≥85% ACS, Fluka™
Magnesium perchlorate
Formula: Cl₂MgO₈
MW: 223.21 g/mol
Melting Pt: 251 (dec.)
Density: 2.21 g/cm³ (18 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011102
CAS Number: 10034-81-8
UN: 1475
ADR: 5.1 II
Merck Index: 13,05702

Order Now

Specification Test Results

Identity (IR) complying
Assay of Mg Min. 10.0 %
Loss on drying (190°C) Max. 8 %
Titrable Acid Max. 0.005 meq/g
Titrable Base Max. 0.025 meq/g
Water absorbing capacity Min. 25 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR